Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGTGACAAAGAGCAAGTCAAG[C/G]CAAGGAAAAAGCTCTCACAAAGAAC
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604942 | |||||||||||||||||||||||||||||
Literature Links: |
FKBP1A-SDCBP2 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FKBP1A-SDCBP2 - FKBP1A-SDCBP2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDCBP2 - syndecan binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199784.1 | 1435 | UTR 3 | NP_001186713.1 | |||
NM_015685.5 | 1435 | UTR 3 | NP_056500.2 | |||
NM_080489.4 | 1435 | UTR 3 | NP_536737.3 |
SNPH - syntaphilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318234.1 | 1435 | Intron | NP_001305163.1 | |||
NM_014723.3 | 1435 | Intron | NP_055538.2 | |||
XM_005260889.3 | 1435 | Intron | XP_005260946.1 | |||
XM_011529402.2 | 1435 | Intron | XP_011527704.1 | |||
XM_011529403.2 | 1435 | Intron | XP_011527705.1 | |||
XM_011529404.2 | 1435 | Intron | XP_011527706.1 |
Set Membership: |
HapMap |