Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGCCCATCTTGGCAGAAGGC[C/T]CAATTACTTCTTGCCTGGTGCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605830 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGFRL1 - fibroblast growth factor receptor-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004356.2 | Intron | NP_001004356.1 | ||||
NM_001004358.1 | Intron | NP_001004358.1 | ||||
NM_021923.3 | Intron | NP_068742.2 | ||||
XM_011513486.1 | Intron | XP_011511788.1 |
Set Membership: |
HapMap |