Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGCCGGGGTGCACACAAGCTG[C/T]GGCGGGGAGGGGCAGGAGCAGAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610303 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAFA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAFA - MAF bZIP transcription factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC3H3 - zinc finger CCCH-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015117.2 | Intron | NP_055932.2 | ||||
XM_006716536.3 | Intron | XP_006716599.2 | ||||
XM_011516943.2 | Intron | XP_011515245.1 | ||||
XM_011516944.2 | Intron | XP_011515246.1 | ||||
XM_017013248.1 | Intron | XP_016868737.1 | ||||
XM_017013249.1 | Intron | XP_016868738.1 |