Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCACCAACACTGCCGAATGTC[A/G]TCTCTTCTCATCTTTATCTCTATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C15orf32 - chromosome 15 open reading frame 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301106.1 | 3629 | Intron | NP_001288035.1 | |||
NM_153040.2 | 3629 | Intron | NP_694585.1 | |||
XM_011521257.2 | 3629 | UTR 5 | XP_011519559.1 |
LOC104613533 - uncharacterized LOC104613533 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST8SIA2 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006011.3 | 3629 | Intron | NP_006002.1 | |||
XM_017022642.1 | 3629 | Intron | XP_016878131.1 | |||
XM_017022643.1 | 3629 | Intron | XP_016878132.1 |