Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCGCTGGAGGGACACTCACCCC[A/G]TCTGCTCCCACGGCAGCACCATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602546 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C15orf32 - chromosome 15 open reading frame 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301106.1 | 4803 | Intron | NP_001288035.1 | |||
NM_153040.2 | 4803 | Intron | NP_694585.1 | |||
XM_011521257.2 | 4803 | UTR 5 | XP_011519559.1 |
LOC104613533 - uncharacterized LOC104613533 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST8SIA2 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006011.3 | 4803 | UTR 3 | NP_006002.1 | |||
XM_017022642.1 | 4803 | UTR 3 | XP_016878131.1 | |||
XM_017022643.1 | 4803 | UTR 3 | XP_016878132.1 |
Set Membership: |
HapMap |