Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606137 MIM: 614058 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGREF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CGREF1 - cell growth regulator with EF-hand domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166239.1 | 672 | Intron | NP_001159711.1 | |||
NM_001166240.1 | 672 | Intron | NP_001159712.1 | |||
NM_001301324.1 | 672 | Silent Mutation | TTA,TTG | L,L 104 | NP_001288253.1 | |
NM_006569.5 | 672 | Intron | NP_006560.3 |
KHK - ketohexokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000221.2 | 672 | UTR 3 | NP_000212.1 | |||
NM_006488.2 | 672 | UTR 3 | NP_006479.1 | |||
XM_005264294.3 | 672 | UTR 3 | XP_005264351.1 | |||
XM_005264296.3 | 672 | UTR 3 | XP_005264353.1 | |||
XM_005264298.3 | 672 | UTR 3 | XP_005264355.1 | |||
XM_006712008.3 | 672 | Intron | XP_006712071.1 | |||
XM_006712009.3 | 672 | Intron | XP_006712072.1 | |||
XM_006712010.3 | 672 | Intron | XP_006712073.1 | |||
XM_006712011.3 | 672 | Intron | XP_006712074.1 | |||
XM_006712012.3 | 672 | Intron | XP_006712075.1 | |||
XM_006712013.3 | 672 | Intron | XP_006712076.1 | |||
XM_006712014.3 | 672 | Intron | XP_006712077.1 | |||
XM_017004060.1 | 672 | UTR 3 | XP_016859549.1 | |||
XM_017004061.1 | 672 | UTR 3 | XP_016859550.1 |
Set Membership: |
HapMap |