Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601513 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGF12 - fibroblast growth factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004113.5 | 1710 | UTR 3 | NP_004104.3 | |||
NM_021032.4 | 1710 | UTR 3 | NP_066360.1 | |||
XM_005247227.2 | 1710 | UTR 3 | XP_005247284.1 | |||
XM_006713538.3 | 1710 | UTR 3 | XP_006713601.1 | |||
XM_017005879.1 | 1710 | UTR 3 | XP_016861368.1 |
Set Membership: |
HapMap |