Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600385 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ADCY7 - adenylate cyclase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114.4 | Intron | NP_001105.1 | ||||
NM_001286057.1 | Intron | NP_001272986.1 | ||||
XM_011522835.2 | Intron | XP_011521137.1 | ||||
XM_011522836.2 | Intron | XP_011521138.1 | ||||
XM_011522837.2 | Intron | XP_011521139.1 | ||||
XM_011522838.2 | Intron | XP_011521140.1 | ||||
XM_011522839.2 | Intron | XP_011521141.1 | ||||
XM_011522840.2 | Intron | XP_011521142.1 | ||||
XM_011522842.2 | Intron | XP_011521144.1 | ||||
XM_017022895.1 | Intron | XP_016878384.1 | ||||
XM_017022896.1 | Intron | XP_016878385.1 | ||||
XM_017022897.1 | Intron | XP_016878386.1 | ||||
XM_017022898.1 | Intron | XP_016878387.1 | ||||
XM_017022899.1 | Intron | XP_016878388.1 |