Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150341 MIM: 607383 MIM: 610477 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LMNB2 - lamin B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032737.3 | 4554 | UTR 3 | NP_116126.3 |
MIR7108 - microRNA 7108 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM13 - translocase of inner mitochondrial membrane 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012458.3 | 4554 | Intron | NP_036590.1 |
TMPRSS9 - transmembrane protease, serine 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |