Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTGGTGGCCACACAAGACACA[C/G]AGCCAGATGCAGGTCTCCTGGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615653 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100132111 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100132111 - uncharacterized LOC100132111 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THEM5 - thioesterase superfamily member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182578.3 | Intron | NP_872384.1 | ||||
XM_011509421.1 | Intron | XP_011507723.1 |
Set Membership: |
Validated |