Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606001 MIM: 608482 | ||||||||||||||||||||||||||||||||
Literature Links: |
IL32 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
IL32 - interleukin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012631.1 | 17 | UTR 5 | NP_001012649.1 | |||
NM_001012632.1 | 17 | Intron | NP_001012650.1 | |||
NM_001012633.1 | 17 | Intron | NP_001012651.1 | |||
NM_001012634.1 | 17 | Intron | NP_001012652.1 | |||
NM_001012635.1 | 17 | UTR 5 | NP_001012653.1 | |||
NM_001012636.1 | 17 | Intron | NP_001012654.1 | |||
NM_001012718.1 | 17 | Intron | NP_001012736.1 | |||
NM_001308078.1 | 17 | Intron | NP_001295007.1 | |||
NM_004221.4 | 17 | Intron | NP_004212.4 |
MMP25 - matrix metallopeptidase 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP25-AS1 - MMP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |