Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGAAAGCCCCCACTGCCTCTC[C/T]CTGTGCCGGCAGGTCAGCGTCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605737 MIM: 612873 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIRC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BIRC7 - baculoviral IAP repeat containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985425 - uncharacterized LOC107985425 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3196 - microRNA 3196 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKAIN4 - Na+/K+ transporting ATPase interacting 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152864.3 | 1187 | Intron | NP_690603.3 | |||
XM_005260192.1 | 1187 | Intron | XP_005260249.1 | |||
XM_005260194.3 | 1187 | Intron | XP_005260251.1 | |||
XM_011528527.1 | 1187 | UTR 3 | XP_011526829.1 | |||
XM_011528528.2 | 1187 | UTR 3 | XP_011526830.1 | |||
XM_011528529.2 | 1187 | UTR 3 | XP_011526831.1 | |||
XM_017027636.1 | 1187 | UTR 3 | XP_016883125.1 | |||
XM_017027637.1 | 1187 | UTR 3 | XP_016883126.1 | |||
XM_017027638.1 | 1187 | Intron | XP_016883127.1 | |||
XM_017027639.1 | 1187 | Intron | XP_016883128.1 |