Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TPRG1 - tumor protein p63 regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198485.3 | Intron | NP_940887.1 | ||||
XM_005247382.4 | Intron | XP_005247439.1 | ||||
XM_005247388.3 | Intron | XP_005247445.1 | ||||
XM_011512732.2 | Intron | XP_011511034.2 | ||||
XM_011512734.2 | Intron | XP_011511036.1 | ||||
XM_011512735.2 | Intron | XP_011511037.1 | ||||
XM_011512736.2 | Intron | XP_011511038.1 | ||||
XM_017006257.1 | Intron | XP_016861746.1 | ||||
XM_017006258.1 | Intron | XP_016861747.1 | ||||
XM_017006259.1 | Intron | XP_016861748.1 | ||||
XM_017006260.1 | Intron | XP_016861749.1 | ||||
XM_017006261.1 | Intron | XP_016861750.1 | ||||
XM_017006262.1 | Intron | XP_016861751.1 | ||||
XM_017006263.1 | Intron | XP_016861752.1 |