Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTGATCTTTTATTTCAGTGATT[C/T]GGAGGACGTTGAAATAACAATAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605228 MIM: 614967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIR1 - corepressor interacting with RBPJ, 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN3 - secernin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193528.1 | Intron | NP_001180457.1 | ||||
NM_024583.4 | Intron | NP_078859.2 | ||||
XM_005246853.2 | Intron | XP_005246910.1 | ||||
XM_005246854.4 | Intron | XP_005246911.1 | ||||
XM_005246855.2 | Intron | XP_005246912.1 | ||||
XM_005246856.2 | Intron | XP_005246913.1 | ||||
XM_005246857.2 | Intron | XP_005246914.1 | ||||
XM_011511839.2 | Intron | XP_011510141.1 | ||||
XM_017004909.1 | Intron | XP_016860398.1 | ||||
XM_017004910.1 | Intron | XP_016860399.1 |