Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGTCACCCTGCTTCTCCCAGC[C/T]GGAGTAGGTGGGGGAGGCCAGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612547 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WBSCR28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WBSCR28 - Williams-Beuren syndrome chromosome region 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182504.3 | 13 | UTR 5 | NP_872310.2 | |||
XM_011515785.2 | 13 | Intron | XP_011514087.1 | |||
XM_017011741.1 | 13 | Intron | XP_016867230.1 |
Set Membership: |
HapMap |