Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTTCAAATTTGTGTAATTTAC[A/C]CCCTGGGAAGATCTGAGTTATTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608427 MIM: 602544 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACRG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PACRG - PARK2 coregulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080378.1 | Intron | NP_001073847.1 | ||||
NM_001080379.1 | Intron | NP_001073848.1 | ||||
NM_152410.2 | Intron | NP_689623.2 | ||||
XM_005266825.4 | Intron | XP_005266882.1 | ||||
XM_005266826.4 | Intron | XP_005266883.1 | ||||
XM_006715344.3 | Intron | XP_006715407.1 | ||||
XM_006715345.3 | Intron | XP_006715408.1 | ||||
XM_011535461.2 | Intron | XP_011533763.1 | ||||
XM_011535462.2 | Intron | XP_011533764.1 | ||||
XM_011535466.1 | Intron | XP_011533768.1 | ||||
XM_011535468.2 | Intron | XP_011533770.1 | ||||
XM_011535469.2 | Intron | XP_011533771.1 | ||||
XM_011535471.1 | Intron | XP_011533773.1 | ||||
XM_017010275.1 | Intron | XP_016865764.1 | ||||
XM_017010276.1 | Intron | XP_016865765.1 | ||||
XM_017010277.1 | Intron | XP_016865766.1 | ||||
XM_017010278.1 | Intron | XP_016865767.1 | ||||
XM_017010279.1 | Intron | XP_016865768.1 | ||||
XM_017010280.1 | Intron | XP_016865769.1 | ||||
XM_017010281.1 | Intron | XP_016865770.1 | ||||
XM_017010282.1 | Intron | XP_016865771.1 |
PARK2 - parkin RBR E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |