Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
79 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605951 MIM: 191164 MIM: 613683 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPM3 - dolichyl-phosphate mannosyltransferase subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA1 - ephrin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004428.2 | 515 | Missense Mutation | GAC,GTC | D,V 159 | NP_004419.2 | |
NM_182685.1 | 515 | Missense Mutation | GAC,GTC | D,V 137 | NP_872626.1 |
SLC50A1 - solute carrier family 50 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |