Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCCTGCACCCCCAACATAAAAC[A/G]TGAAGCCAGGACACATTGACAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286389.1 | Intron | NP_001273318.1 | ||||
NM_001286390.1 | Intron | NP_001273319.1 | ||||
XM_011536745.2 | Intron | XP_011535047.1 | ||||
XM_011536746.2 | Intron | XP_011535048.1 | ||||
XM_011536750.2 | Intron | XP_011535052.1 |