Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCTGGGCAGACGACTCTGAACA[C/T]GGCTTCATATTGGCCTTCCATGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNIH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNIH3 - cornichon family AMPA receptor auxiliary protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322302.1 | Intron | NP_001309231.1 | ||||
NM_001322303.1 | Intron | NP_001309232.1 | ||||
NM_001322304.1 | Intron | NP_001309233.1 | ||||
NM_001322305.1 | Intron | NP_001309234.1 | ||||
NM_152495.1 | Intron | NP_689708.1 |
WDR26 - WD repeat domain 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |