Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603145 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPFIA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPFIA4 - PTPRF interacting protein alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304331.1 | Intron | NP_001291260.1 | ||||
NM_001304332.1 | Intron | NP_001291261.1 | ||||
XM_006711586.2 | Intron | XP_006711649.1 | ||||
XM_011510070.2 | Intron | XP_011508372.1 | ||||
XM_017002606.1 | Intron | XP_016858095.1 | ||||
XM_017002607.1 | Intron | XP_016858096.1 | ||||
XM_017002608.1 | Intron | XP_016858097.1 | ||||
XM_017002609.1 | Intron | XP_016858098.1 | ||||
XM_017002610.1 | Intron | XP_016858099.1 |
TMEM183A - transmembrane protein 183A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |