Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
72 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611673 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SWT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105518.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | NP_001098988.1 | |
NM_017673.6 | 291 | Silent Mutation | TCA,TCG | S,S 42 | NP_060143.4 | |
XM_005245271.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_005245328.1 | |
XM_005245273.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_005245330.1 | |
XM_011509657.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_011507959.1 | |
XM_017001527.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_016857016.1 | |
XM_017001528.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_016857017.1 | |
XM_017001529.1 | 291 | UTR 5 | XP_016857018.1 | |||
XM_017001530.1 | 291 | Silent Mutation | TCA,TCG | S,S 42 | XP_016857019.1 |
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |