Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGAAAACACTGAACTAGAGAAC[A/C]GGAAGAGCTGGACACCTATTTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616169 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOMM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TOMM5 - translocase of outer mitochondrial membrane 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001790.2 | Intron | NP_001001790.1 | ||||
NM_001134484.1 | Intron | NP_001127956.1 | ||||
NM_001134485.1 | Intron | NP_001127957.1 |
Set Membership: |
HapMap |