Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602245 MIM: 613569 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTPBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTPBP1 - GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004286.4 | Intron | NP_004277.2 | ||||
XM_011530537.2 | Intron | XP_011528839.1 | ||||
XM_017029099.1 | Intron | XP_016884588.1 | ||||
XM_017029100.1 | Intron | XP_016884589.1 | ||||
XM_017029101.1 | Intron | XP_016884590.1 | ||||
XM_017029102.1 | Intron | XP_016884591.1 | ||||
XM_017029103.1 | Intron | XP_016884592.1 |
SUN2 - Sad1 and UNC84 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199579.1 | Intron | NP_001186508.1 | ||||
NM_001199580.1 | Intron | NP_001186509.1 | ||||
NM_015374.2 | Intron | NP_056189.1 | ||||
XM_011530104.2 | Intron | XP_011528406.1 | ||||
XM_011530105.2 | Intron | XP_011528407.1 | ||||
XM_017028748.1 | Intron | XP_016884237.1 |
Set Membership: |
HapMap |