Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606991 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDHR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDHR4 - cadherin related family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007540.3 | Intron | NP_001007541.2 | ||||
XM_011533700.2 | Intron | XP_011532002.1 | ||||
XM_011533701.2 | Intron | XP_011532003.1 | ||||
XM_017006366.1 | Intron | XP_016861855.1 | ||||
XM_017006367.1 | Intron | XP_016861856.1 | ||||
XM_017006368.1 | Intron | XP_016861857.1 | ||||
XM_017006369.1 | Intron | XP_016861858.1 | ||||
XM_017006370.1 | Intron | XP_016861859.1 | ||||
XM_017006371.1 | Intron | XP_016861860.1 |
FAM212A - family with sequence similarity 212 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IP6K1 - inositol hexakisphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |