Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606696 MIM: 603473 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KATNA1 - katanin catalytic subunit A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270519.1 | 6767 | Intron | NP_001257448.1 | |||
NM_004690.3 | 6767 | UTR 3 | NP_004681.1 | |||
XM_006715603.3 | 6767 | Intron | XP_006715666.1 | |||
XM_011536252.2 | 6767 | Intron | XP_011534554.1 | |||
XM_017011474.1 | 6767 | UTR 3 | XP_016866963.1 | |||
XM_017011475.1 | 6767 | UTR 3 | XP_016866964.1 | |||
XM_017011476.1 | 6767 | UTR 3 | XP_016866965.1 | |||
XM_017011477.1 | 6767 | Intron | XP_016866966.1 | |||
XM_017011478.1 | 6767 | UTR 3 | XP_016866967.1 | |||
XM_017011479.1 | 6767 | Intron | XP_016866968.1 | |||
XM_017011480.1 | 6767 | UTR 3 | XP_016866969.1 | |||
XM_017011481.1 | 6767 | Intron | XP_016866970.1 | |||
XM_017011482.1 | 6767 | Intron | XP_016866971.1 |
Set Membership: |
HapMap |