Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAATCTTGCTCCAAGTGGAACAC[C/T]GGTTGTCTGTATGAGAGCCAACGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
RAD54L2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RAD54L2 - RAD54-like 2 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322253.1 | Intron | NP_001309182.1 | ||||
NM_001322256.1 | Intron | NP_001309185.1 | ||||
NM_001322257.1 | Intron | NP_001309186.1 | ||||
NM_015106.3 | Intron | NP_055921.2 | ||||
XM_006713062.3 | Intron | XP_006713125.1 | ||||
XM_017005987.1 | Intron | XP_016861476.1 | ||||
XM_017005988.1 | Intron | XP_016861477.1 |