Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCTGGGGAGGGTGGGCAGCGGG[T/C]GGCACTGGGCCCAGGGAGGGGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602859 MIM: 612836 | ||||||||||||||||||||
Literature Links: |
PEX10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEX10 - peroxisomal biogenesis factor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCH2 - phospholipase C eta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303012.1 | Intron | NP_001289941.1 | ||||
NM_001303013.1 | Intron | NP_001289942.1 | ||||
NM_014638.3 | Intron | NP_055453.2 | ||||
XM_011542455.2 | Intron | XP_011540757.1 | ||||
XM_011542456.2 | Intron | XP_011540758.1 | ||||
XM_017002870.1 | Intron | XP_016858359.1 | ||||
XM_017002871.1 | Intron | XP_016858360.1 | ||||
XM_017002872.1 | Intron | XP_016858361.1 | ||||
XM_017002873.1 | Intron | XP_016858362.1 | ||||
XM_017002874.1 | Intron | XP_016858363.1 |