Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf26 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C7orf26 - chromosome 7 open reading frame 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF853 - zinc finger protein 853 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017560.1 | 452 | Missense Mutation | CAA,CGA | Q,R 30 | NP_060030.1 | |
XM_011515438.2 | 452 | Missense Mutation | CAA,CGA | Q,R 30 | XP_011513740.1 | |
XM_011515439.2 | 452 | Missense Mutation | CAA,CGA | Q,R 15 | XP_011513741.1 | |
XM_017012371.1 | 452 | Missense Mutation | CAA,CGA | Q,R 15 | XP_016867860.1 |
Set Membership: |
HapMap Validated |