Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf229 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf229 - chromosome 6 open reading frame 229 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM65B - family with sequence similarity 65 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286445.1 | 4314 | UTR 3 | NP_001273374.1 | |||
NM_001286446.1 | 4314 | Intron | NP_001273375.1 | |||
NM_001286447.1 | 4314 | Intron | NP_001273376.1 | |||
NM_014722.3 | 4314 | UTR 3 | NP_055537.2 | |||
NM_015864.3 | 4314 | Intron | NP_056948.2 | |||
XM_006715275.2 | 4314 | UTR 3 | XP_006715338.1 | |||
XM_006715277.1 | 4314 | UTR 3 | XP_006715340.1 | |||
XM_006715279.2 | 4314 | UTR 3 | XP_006715342.1 | |||
XM_006715281.3 | 4314 | Intron | XP_006715344.1 | |||
XM_011515007.1 | 4314 | UTR 3 | XP_011513309.1 | |||
XM_011515008.2 | 4314 | UTR 3 | XP_011513310.1 | |||
XM_011515009.1 | 4314 | UTR 3 | XP_011513311.1 | |||
XM_011515010.2 | 4314 | UTR 3 | XP_011513312.1 | |||
XM_011515012.1 | 4314 | UTR 3 | XP_011513314.1 | |||
XM_017011523.1 | 4314 | UTR 3 | XP_016867012.1 | |||
XM_017011524.1 | 4314 | Intron | XP_016867013.1 | |||
XM_017011525.1 | 4314 | Intron | XP_016867014.1 | |||
XM_017011526.1 | 4314 | Intron | XP_016867015.1 | |||
XM_017011527.1 | 4314 | Intron | XP_016867016.1 |