Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTAACAAGGTAAAGGACAGAGT[A/G]CGGTGCAGAGGTCAGGGTACAGAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154550 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM219B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM219B - family with sequence similarity 219 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPI - mannose phosphate isomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289155.1 | Intron | NP_001276084.1 | ||||
NM_001289156.1 | Intron | NP_001276085.1 | ||||
NM_001289157.1 | Intron | NP_001276086.1 | ||||
NM_002435.2 | Intron | NP_002426.1 | ||||
XM_011521592.1 | Intron | XP_011519894.1 | ||||
XM_011521593.2 | Intron | XP_011519895.1 | ||||
XM_017022207.1 | Intron | XP_016877696.1 | ||||
XM_017022208.1 | Intron | XP_016877697.1 | ||||
XM_017022209.1 | Intron | XP_016877698.1 |
Set Membership: |
HapMap |