Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATGTTCTGAAAATTCATCTTT[A/C]ACATTTCTTTGATAATTTGTACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194552 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF79 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286696.1 | Intron | NP_001273625.1 | ||||
NM_001286697.1 | Intron | NP_001273626.1 | ||||
NM_001286698.1 | Intron | NP_001273627.1 | ||||
NM_001322260.1 | Intron | NP_001309189.1 | ||||
NM_007135.2 | Intron | NP_009066.2 | ||||
XM_006717279.3 | Intron | XP_006717342.1 |
Set Membership: |
HapMap |