Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606590 MIM: 180073 | ||||||||||||||||||||
Literature Links: |
NPLOC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPLOC4 - NPL4 homolog, ubiquitin recognition factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290212.1 | 694 | Missense Mutation | ACA,GCA | T,A 156 | NP_001277141.1 | |
NM_031945.4 | 694 | Missense Mutation | ACA,GCA | T,A 118 | NP_114151.3 |