Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATCGCCTCAGTGAAGGTCACA[C/T]TGCAGCACTATTGATTTCTAAAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606738 | |||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL10 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
OSBPL10 - oxysterol binding protein like 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF860 - zinc finger protein 860 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137674.2 | 522 | UTR 5 | NP_001131146.2 | |||
XM_017006299.1 | 522 | Intron | XP_016861788.1 |