Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAGTCTCACCTCCGTGCAACC[A/C]GAAGGAGGATTGGAATATGGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606101 MIM: 616838 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRE3 - adhesion G protein-coupled receptor E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289158.1 | Intron | NP_001276087.1 | ||||
NM_001289159.1 | Intron | NP_001276088.1 | ||||
NM_032571.4 | Intron | NP_115960.2 | ||||
XM_011528374.2 | Intron | XP_011526676.1 | ||||
XM_017027383.1 | Intron | XP_016882872.1 |
CLEC17A - C-type lectin domain family 17 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204118.1 | Intron | NP_001191047.1 | ||||
NM_207390.3 | Intron | NP_997273.3 | ||||
XM_017026785.1 | Intron | XP_016882274.1 | ||||
XM_017026786.1 | Intron | XP_016882275.1 | ||||
XM_017026787.1 | Intron | XP_016882276.1 | ||||
XM_017026788.1 | Intron | XP_016882277.1 | ||||
XM_017026789.1 | Intron | XP_016882278.1 | ||||
XM_017026790.1 | Intron | XP_016882279.1 | ||||
XM_017026791.1 | Intron | XP_016882280.1 | ||||
XM_017026792.1 | Intron | XP_016882281.1 | ||||
XM_017026793.1 | Intron | XP_016882282.1 | ||||
XM_017026794.1 | Intron | XP_016882283.1 |
Set Membership: |
HapMap |