Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGCCTCGTTGTACATCAGAGAC[A/G]GAGCATTTTACACCTTGAAGACGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191740 MIM: 606435 MIM: 606428 MIM: 606429 MIM: 606430 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100286922 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
UGT1A1*6,c.211G>A UGT1A1*6,g.211G>A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100286922 - DnaJ heat shock protein family (Hsp40) member B3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UGT1A1 - UDP glucuronosyltransferase family 1 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000463.2 | 226 | Missense Mutation | AGA,GGA | R,G 71 | NP_000454.1 |
UGT1A10 - UDP glucuronosyltransferase family 1 member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019075.2 | 226 | Intron | NP_061948.1 |
UGT1A3 - UDP glucuronosyltransferase family 1 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019093.2 | 226 | Intron | NP_061966.1 |
UGT1A4 - UDP glucuronosyltransferase family 1 member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007120.2 | 226 | Intron | NP_009051.1 |
UGT1A5 - UDP glucuronosyltransferase family 1 member A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019078.1 | 226 | Intron | NP_061951.1 |
UGT1A6 - UDP glucuronosyltransferase family 1 member A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001072.3 | 226 | Intron | NP_001063.2 | |||
NM_205862.1 | 226 | Intron | NP_995584.1 |
UGT1A7 - UDP glucuronosyltransferase family 1 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019077.2 | 226 | Intron | NP_061950.2 |
UGT1A8 - UDP glucuronosyltransferase family 1 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019076.4 | 226 | Intron | NP_061949.3 |
UGT1A9 - UDP glucuronosyltransferase family 1 member A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021027.2 | 226 | Intron | NP_066307.1 |
Set Membership: |
HapMap DME Validated Inventoried |