Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
87 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYSM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYSM1 - Myb like, SWIRM and MPN domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085487.2 | 2476 | Missense Mutation | AAA,GAA | K,E 825 | NP_001078956.1 | |
XM_006710314.3 | 2476 | Missense Mutation | AAA,GAA | K,E 824 | XP_006710377.1 | |
XM_011540573.2 | 2476 | Missense Mutation | AAA,GAA | K,E 758 | XP_011538875.1 | |
XM_011540574.2 | 2476 | Missense Mutation | AAA,GAA | K,E 736 | XP_011538876.1 | |
XM_017000195.1 | 2476 | Missense Mutation | AAA,GAA | K,E 572 | XP_016855684.1 | |
XM_017000196.1 | 2476 | Missense Mutation | AAA,GAA | K,E 572 | XP_016855685.1 | |
XM_017000197.1 | 2476 | Missense Mutation | AAA,GAA | K,E 572 | XP_016855686.1 |
Set Membership: |
HapMap |