Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTCCACAGAGACTTGTCATTC[A/C]GCATGCCAGAGGTGAAATGACCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606890 | ||||||||||||||||||||
Literature Links: |
GALC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALC - galactosylceramidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000153.3 | 2323 | Missense Mutation | CGG,CTG | R,L 645 | NP_000144.2 | |
NM_001201401.1 | 2323 | Missense Mutation | CGG,CTG | R,L 622 | NP_001188330.1 | |
NM_001201402.1 | 2323 | Missense Mutation | CGG,CTG | R,L 619 | NP_001188331.1 | |
XM_011536618.2 | 2323 | Missense Mutation | CGG,CTG | R,L 589 | XP_011534920.1 |