Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATTAACTGCTTAAAACAGTGC[A/G]CAGGCATGTGGGCTGGGTGCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612428 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100291105 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100291105 - uncharacterized LOC100291105 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM38 - RNA binding motif protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291780.1 | Intron | NP_001278709.1 | ||||
NM_017495.5 | Intron | NP_059965.2 | ||||
NM_183425.2 | Intron | NP_906270.1 | ||||
XM_005260446.3 | Intron | XP_005260503.1 | ||||
XM_011528885.2 | Intron | XP_011527187.1 |