Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612779 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPYD - dihydropyrimidine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000110.3 | 4193 | UTR 3 | NP_000101.2 | |||
NM_001160301.1 | 4193 | Intron | NP_001153773.1 | |||
XM_005270562.3 | 4193 | UTR 3 | XP_005270619.2 | |||
XM_006710397.3 | 4193 | Intron | XP_006710460.1 | |||
XM_017000507.1 | 4193 | UTR 3 | XP_016855996.1 | |||
XM_017000508.1 | 4193 | UTR 3 | XP_016855997.1 | |||
XM_017000509.1 | 4193 | UTR 3 | XP_016855998.1 | |||
XM_017000510.1 | 4193 | UTR 3 | XP_016855999.1 |
Set Membership: |
HapMap |