Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138430 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPD2 - glycerol-3-phosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000408.4 | Intron | NP_000399.3 | ||||
NM_001083112.2 | Intron | NP_001076581.2 | ||||
XM_005246469.2 | Intron | XP_005246526.1 | ||||
XM_011510977.2 | Intron | XP_011509279.1 | ||||
XM_011510978.2 | Intron | XP_011509280.1 | ||||
XM_011510980.2 | Intron | XP_011509282.1 | ||||
XM_017003830.1 | Intron | XP_016859319.1 | ||||
XM_017003831.1 | Intron | XP_016859320.1 |
Set Membership: |
HapMap |