Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCTGTTGGGGTTTAGCGAACGG[T/C]CAGCAATCACAACAGCGGTACTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 107300 | ||||||||||||||||||||
Literature Links: |
SERPINC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINC1 - serpin family C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000488.3 | 1390 | Missense Mutation | GAC,GGC | D,G 424 | NP_000479.1 | |
XM_005245198.3 | 1390 | Missense Mutation | GAC,GGC | D,G 376 | XP_005245255.1 |