Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601604 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL12RB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IL12RB1 - interleukin 12 receptor subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290023.1 | 2955 | UTR 3 | NP_001276952.1 | |||
NM_001290024.1 | 2955 | UTR 3 | NP_001276953.1 | |||
NM_005535.2 | 2955 | UTR 3 | NP_005526.1 | |||
NM_153701.2 | 2955 | Intron | NP_714912.1 | |||
XM_006722741.3 | 2955 | UTR 3 | XP_006722804.2 | |||
XM_011527966.2 | 2955 | UTR 3 | XP_011526268.1 | |||
XM_011527967.2 | 2955 | UTR 3 | XP_011526269.1 | |||
XM_011527968.2 | 2955 | UTR 3 | XP_011526270.1 | |||
XM_011527969.2 | 2955 | UTR 3 | XP_011526271.1 | |||
XM_011527970.2 | 2955 | UTR 3 | XP_011526272.1 | |||
XM_011527971.2 | 2955 | UTR 3 | XP_011526273.1 | |||
XM_011527972.2 | 2955 | UTR 3 | XP_011526274.1 | |||
XM_011527973.2 | 2955 | UTR 3 | XP_011526275.1 | |||
XM_011527974.2 | 2955 | UTR 3 | XP_011526276.1 | |||
XM_011527975.2 | 2955 | UTR 3 | XP_011526277.1 | |||
XM_011527976.2 | 2955 | Intron | XP_011526278.1 | |||
XM_011527977.2 | 2955 | Intron | XP_011526279.1 | |||
XM_017026762.1 | 2955 | UTR 3 | XP_016882251.1 |
Set Membership: |
HapMap Validated |