Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603261 MIM: 602176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP4K2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIP4K2B - phosphatidylinositol-5-phosphate 4-kinase type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003559.4 | 1536 | Silent Mutation | NP_003550.1 | |||
XM_011525326.2 | 1536 | Silent Mutation | XP_011523628.1 | |||
XM_011525327.2 | 1536 | Silent Mutation | XP_011523629.1 | |||
XM_011525329.2 | 1536 | Intron | XP_011523631.1 | |||
XM_011525330.2 | 1536 | Silent Mutation | XP_011523632.1 | |||
XM_017025196.1 | 1536 | Silent Mutation | XP_016880685.1 | |||
XM_017025197.1 | 1536 | Silent Mutation | XP_016880686.1 | |||
XM_017025198.1 | 1536 | Silent Mutation | XP_016880687.1 | |||
XM_017025199.1 | 1536 | Silent Mutation | XP_016880688.1 |
PSMB3 - proteasome subunit beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |