Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PHF14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF14 - PHD finger protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014660.3 | 487 | UTR 5 | NP_055475.2 | |||
XM_017012811.1 | 487 | UTR 5 | XP_016868300.1 | |||
XM_017012812.1 | 487 | UTR 5 | XP_016868301.1 | |||
XM_017012813.1 | 487 | UTR 5 | XP_016868302.1 | |||
XM_017012814.1 | 487 | UTR 5 | XP_016868303.1 | |||
XM_017012815.1 | 487 | UTR 5 | XP_016868304.1 | |||
XM_017012816.1 | 487 | UTR 5 | XP_016868305.1 | |||
XM_017012817.1 | 487 | UTR 5 | XP_016868306.1 |