Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGAGGTTGGCAATTTAAGAGTT[C/G]TTATTAAAATAATAATTTACCTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609232 | ||||||||||||||||||||
Literature Links: |
LOC102467212 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102467212 - uncharacterized LOC102467212 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT12 - nudix hydrolase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300741.1 | 3302 | UTR 3 | NP_001287670.1 | |||
NM_031438.3 | 3302 | UTR 3 | NP_113626.1 | |||
XM_005272095.1 | 3302 | UTR 3 | XP_005272152.1 | |||
XM_005272097.3 | 3302 | UTR 3 | XP_005272154.1 |