Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608092 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CBR4 - carbonyl reductase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032783.4 | Intron | NP_116172.2 | ||||
XM_005263315.2 | Intron | XP_005263372.1 | ||||
XM_005263316.3 | Intron | XP_005263373.1 | ||||
XM_006714391.1 | Intron | XP_006714454.1 | ||||
XM_006714392.3 | Intron | XP_006714455.1 | ||||
XM_011532385.1 | Intron | XP_011530687.1 | ||||
XM_011532386.2 | Intron | XP_011530688.1 | ||||
XM_017008782.1 | Intron | XP_016864271.1 | ||||
XM_017008783.1 | Intron | XP_016864272.1 |
PALLD - palladin, cytoskeletal associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166108.1 | Intron | NP_001159580.1 | ||||
NM_001166109.1 | Intron | NP_001159581.1 | ||||
NM_001166110.1 | Intron | NP_001159582.1 | ||||
NM_016081.3 | Intron | NP_057165.3 | ||||
XM_005262861.4 | Intron | XP_005262918.1 | ||||
XM_005262866.2 | Intron | XP_005262923.1 | ||||
XM_011531768.2 | Intron | XP_011530070.1 | ||||
XM_011531769.2 | Intron | XP_011530071.1 | ||||
XM_011531770.2 | Intron | XP_011530072.1 | ||||
XM_011531771.2 | Intron | XP_011530073.1 | ||||
XM_011531772.2 | Intron | XP_011530074.1 | ||||
XM_011531773.1 | Intron | XP_011530075.1 | ||||
XM_011531774.1 | Intron | XP_011530076.1 | ||||
XM_011531775.1 | Intron | XP_011530077.1 | ||||
XM_017007910.1 | Intron | XP_016863399.1 | ||||
XM_017007911.1 | Intron | XP_016863400.1 |
Set Membership: |
HapMap Validated |