Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAACAAAAATTGAAATATAAGT[C/T]AGCGATTCTAGAGACCATGCTGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606080 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHIA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHIA - chitinase, acidic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040623.2 | Intron | NP_001035713.1 | ||||
NM_001258001.1 | Intron | NP_001244930.1 | ||||
NM_001258002.1 | Intron | NP_001244931.1 | ||||
NM_001258003.1 | Intron | NP_001244932.1 | ||||
NM_001258004.1 | Intron | NP_001244933.1 | ||||
NM_001258005.1 | Intron | NP_001244934.1 | ||||
NM_021797.3 | Intron | NP_068569.2 | ||||
NM_201653.3 | Intron | NP_970615.2 | ||||
XM_006710577.3 | Intron | XP_006710640.1 | ||||
XM_017001047.1 | Intron | XP_016856536.1 | ||||
XM_017001048.1 | Intron | XP_016856537.1 |
CHIAP2 - chitinase, acidic pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |