Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||
Literature Links: |
NEK10 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
Set Membership: |
Validated |