Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCAGAAGCTTGATAGGAGCCAGG[C/T]TTTGTGCTTTTCGCATGTCACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610526 MIM: 616796 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5C1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NT5C1B - 5'-nucleotidase, cytosolic IB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NT5C1B-RDH14 - NT5C1B-RDH14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199103.1 | Intron | NP_001186032.1 | ||||
NM_001199104.1 | Intron | NP_001186033.1 |
RDH14 - retinol dehydrogenase 14 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020905.3 | Intron | NP_065956.1 |
Set Membership: |
HapMap Validated |