Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGGAAGAAAGAGAATAAACTAA[C/G]TATAAATTATCAGTTCTGTTAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP28 - Rho GTPase activating protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010000.2 | Intron | NP_001010000.1 | ||||
XM_005258144.1 | Intron | XP_005258201.2 | ||||
XM_005258145.1 | Intron | XP_005258202.2 | ||||
XM_005258146.2 | Intron | XP_005258203.1 |
LOC101927168 - uncharacterized LOC101927168 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |